Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126079.1 | NP_001119551.1 | 2 | 98 | ||
XM_006254269.2 | XP_006254331.1 | 2 | 1456 |