Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108427.1 | NP_001101897.1 | 3 | 270 | ||
XM_006254598.1 | XP_006254660.1 | 3 | 303 | |||
GenBank mRNA | FQ227006.1 | - | 742 | |||
FQ234841.1 | - | 743 |