Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306144.1 | NP_001293073.1 | 12 | 1572 | ||
NM_003828.3 | NP_003819.1 | 11 | 1548 | |||
XM_005274765.3 | XP_005274822.1 | 14 | 1612 | |||
XM_005274766.3 | XP_005274823.1 | 13 | 1560 | |||
XM_005274767.3 | XP_005274824.1 | 12 | 1579 | |||
XM_006724855.3 | XP_006724918.1 | 11 | 1548 | |||
XM_011531209.2 | XP_011529511.1 | 11 | 1308 | |||
XM_017029920.1 | XP_016885409.1 | 14 | 1612 | |||
XM_017029921.1 | XP_016885410.1 | 12 | 1343 | |||
XM_017029922.1 | XP_016885411.1 | 12 | 1355 | |||
XM_017029923.1 | XP_016885412.1 | 9 | 1135 | |||
GenBank mRNA | AF057354.1 | - | 11 | 1372 | ||
AJ224979.1 | - | 11 | 1374 | |||
AK001816.1 | - | 8 | 891 | |||
AK023451.1 | - | 2 | 1521 | |||
AK226109.1 | - | 12 | 1526 | |||
AK295738.1 | - | 12 | 1367 | |||
U58032.1 | - | 8 | 1153 |