Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004276.1 | NP_001004276.1 | 3 | 210 | ||
XM_006254697.2 | XP_006254759.1 | 3 | 274 | |||
GenBank mRNA | BC079373.1 | - | 3 | 210 | 7110105 |