Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108441.1 | NP_001101911.1 | 2 | 853 | ||
XM_006255282.2 | XP_006255344.1 | 2 | 820 | |||
GenBank mRNA | BC159430.1 | - | 2 | 716 | 9089687 | |
FQ227962.1 | - | 2 | 775 |