Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003833.4 | NP_003824.2 | 8 | 1314 | ||
NM_030590.3 | NP_085080.1 | 6 | 1133 | |||
NM_030592.3 | NP_085095.1 | 5 | 980 | |||
XM_005260597.1 | XP_005260654.1 | 7 | 1495 | |||
XM_017028113.1 | XP_016883602.1 | 8 | 1314 | |||
XM_017028114.1 | XP_016883603.1 | 6 | 1372 | |||
XM_017028115.1 | XP_016883604.1 | 5 | 1249 | |||
GenBank mRNA | AJ007581.1 | - | 6 | 1078 | ||
AK027323.1 | - | 2 | 170 | |||
AK074495.1 | - | 4 | 512 | |||
AK074593.1 | - | 9 | 1314 | |||
AK074595.1 | - | 6 | 1050 | |||
AK074597.1 | - | 6 | 1135 | |||
BC131763.1 | - | 6 | 1082 | 40121434 | ||
BC141811.1 | - | 6 | 1121 | 40147663 | ||
BC143625.1 | - | 6 | 1041 | |||
BC151219.1 | - | 7 | 1209 | 40148522 |