Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014153.1 | NP_001014175.1 | 5 | 518 | ||
GenBank mRNA | BC085121.1 | - | 5 | 518 | 7115238 | |
FQ211965.1 | - | 561 | ||||
FQ212485.1 | - | 485 |