Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271031.1 | NP_001257960.1 | 3 | 419 | ||
GenBank mRNA | BC089891.1 | - | 3 | 419 |