Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108454.1 | NP_001101924.1 | 11 | 1332 | ||
XM_006255774.2 | XP_006255836.1 | 14 | 1595 | |||
XM_008772632.1 | XP_008770854.1 | 14 | 1979 | |||
GenBank mRNA | AM747283.1 | - | 14 | 1427 |