Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009704.1 | NP_001009704.1 | 3 | 1110 | ||
XM_008772638.1 | XP_008770860.1 | 3 | 1134 | |||
XM_008772639.1 | XP_008770861.1 | 3 | 1134 | |||
GenBank mRNA | AY043227.1 | - | 3 | 1110 |