Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108466.1 | NP_001101936.1 | 1, 2 | 440 | ||
XM_006228268.2 | XP_006228330.1 | 2, 3 | 390 | |||
XM_006228269.2 | XP_006228331.1 | 2, 3 | 387 | |||
XM_006228270.1 | XP_006228332.1 | 2, 3 | 341 | |||
XM_006228271.1 | XP_006228333.1 | 2, 3 | 338 | |||
XM_006228272.2 | XP_006228334.1 | 2, 3 | 694 | |||
XM_006228273.2 | XP_006228335.1 | 2, 3 | 691 |