Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108471.1 | NP_001101941.1 | 6 | 1050 | ||
GenBank mRNA | FQ209516.1 | - | 7 | 643 |