Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025131.1 | NP_001020302.1 | 3, 4 | 503 | ||
GenBank mRNA | D14989.1 | - | 3, 4 | 503 | ||
FQ209413.1 | - | 3, 4 | 516 | |||
FQ218086.1 | - | 3, 4 | 490 | |||
FQ219053.1 | - | 3, 4 | 516 | |||
FQ219201.1 | - | 3, 4 | 516 | |||
FQ219357.1 | - | 3, 4 | 489 | |||
FQ219375.1 | - | 3, 4 | 527 | |||
FQ219766.1 | - | 3, 4 | 515 |