Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108477.1 | NP_001101947.1 | 2 | 460 | ||
XM_008758955.1 | XP_008757177.1 | 2 | 452 | |||
XM_008758956.1 | XP_008757178.1 | 2 | 354 | |||
XM_008758957.1 | XP_008757179.1 | 2 | 136 | |||
GenBank mRNA | FQ222753.1 | - | 2 | 473 |