Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108477.1 | NP_001101947.1 | 2, 3 | 506 | ||
GenBank mRNA | FQ222753.1 | - | 2, 3 | 519 |