Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008775.1 | NP_001008775.1 | 4, 5 | 238 | ||
XM_006228776.2 | XP_006228838.1 | 3, 4 | 255 | |||
XM_006228777.1 | XP_006228839.1 | 2, 3 | 139 | |||
XM_006228778.2 | XP_006228840.1 | 3, 4 | 255 | |||
XM_006228779.2 | XP_006228841.1 | 3, 4 | 255 | |||
GenBank mRNA | AY325237.1 | - | 300 | |||
AY325238.1 | - | 4, 5 | 238 |