Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008775.1 | NP_001008775.1 | 3 | 151 | ||
XM_006228776.2 | XP_006228838.1 | 2 | 168 | |||
XM_006228777.1 | XP_006228839.1 | 1 | 52 | |||
XM_006228778.2 | XP_006228840.1 | 2 | 168 | |||
XM_006228779.2 | XP_006228841.1 | 2 | 168 | |||
GenBank mRNA | AY325237.1 | - | 149 | |||
AY325238.1 | - | 3 | 151 |