Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173357.1 | NP_001166828.1 | 9 | 1296 | ||
GenBank mRNA | BC166816.1 | - | 9 | 1296 | 7462729 |