Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108483.1 | NP_001101953.1 | 8 | 727 | ||
XM_006229101.2 | XP_006229163.1 | 7 | 671 | |||
XM_006229102.2 | XP_006229164.1 | 7 | 593 | |||
GenBank mRNA | BC167051.1 | - | 7 | 509 | 9089731 |