Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012168.1 | NP_001012168.1 | 10 | 1457 | ||
XM_006229103.2 | XP_006229165.1 | 13 | 1653 | |||
XM_006229105.2 | XP_006229167.1 | 13 | 1631 | |||
XM_006229106.2 | XP_006229168.1 | 13 | 1660 | |||
XM_006229107.2 | XP_006229169.1 | 10 | 1357 | |||
XM_008759398.1 | XP_008757620.1 | 10 | 1755 | |||
GenBank mRNA | BC084696.1 | - | 10 | 1457 | 7115051 |