Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nomo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108484.1 | NP_001101954.1 | 22, 23 | 2685 | ||
XM_006229108.2 | XP_006229170.1 | 22, 23 | 2700 |