Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108490.1 | NP_001101960.1 | 6 | 577 | ||
XM_006229441.2 | XP_006229503.1 | 5 | 459 | |||
XM_008759543.1 | XP_008757765.1 | 4 | 839 | |||
XM_008759544.1 | XP_008757766.1 | 5 | 431 |