Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759657.1 | XP_008757879.1 | 2, 3 | 645 | ||
XM_341883.6 | XP_341884.4 | 2, 3 | 645 |