Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1310712 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229802.2 | XP_006229864.1 | 22 | 9611 | ||
XM_006229804.2 | XP_006229866.1 | 21 | 9569 | |||
XM_006229805.2 | XP_006229867.1 | 21 | 9569 | |||
XM_006229807.2 | XP_006229869.1 | 21 | 9461 | |||
XM_006229808.2 | XP_006229870.1 | 21 | 9458 | |||
XM_006229809.2 | XP_006229871.1 | 20 | 9413 | |||
XM_006229810.2 | XP_006229872.1 | 19 | 9248 | |||
XM_006229811.2 | XP_006229873.1 | 21 | 9131 | |||
XM_006229813.2 | XP_006229875.1 | 20 | 9416 | |||
GenBank mRNA | AY724538.1 | - | 4 | 1224 |