Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184795.1 | NP_001171724.1 | 4 | 673 | ||
NM_003846.2 | NP_003837.1 | 4 | 867 | |||
NR_073491.1 | 4 | 863 | ||||
NR_073492.1 | 4 | 857 | ||||
NR_073493.2 | 4 | 1079 | ||||
GenBank mRNA | AB018080.1 | - | 4 | 649 | ||
AF093670.1 | - | 4 | 647 | |||
AK023991.1 | - | 4 | 631 | |||
AK094173.1 | - | 1 | 1467 | |||
AK301983.1 | - | 4 | 633 | |||
BC011963.1 | - | 4 | 647 | 3866690 | ||
CR542047.1 | - | 4 | 631 |