Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108502.2 | NP_001101972.2 | 11 | 1480 | ||
XM_006230127.2 | XP_006230189.1 | 12 | 1633 | |||
XM_006230128.2 | XP_006230190.1 | 11 | 1549 | |||
XM_006230129.2 | XP_006230191.1 | 11 | 1396 | |||
XM_008759771.1 | XP_008757993.1 | 12 | 1549 | |||
XM_008759772.1 | XP_008757994.1 | 11 | 1525 | |||
XM_008759773.1 | XP_008757995.1 | 12 | 1630 | |||
GenBank mRNA | BC168985.1 | - | 11 | 1480 | 7323876 | |
FQ218416.1 | - | 7 | 673 |