Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ears2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760356.1 | XP_008758578.1 | 1 | 1171 |
Gene Symbol: Ears2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159493.1 | NP_001152965.1 | 6 | 1238 |