Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB015594.1 | - | 3 | 504 | ||
AF093668.1 | - | 3 | 466 | |||
AK297442.1 | - | 2 | 293 | |||
AK300978.1 | - | 3 | 346 | |||
AK313318.1 | - | 3 | 475 | |||
BC009697.1 | - | 3 | 455 | 3878406 | ||
BC101164.2 | - | 3 | 415 | 40015908 | ||
CR542046.1 | - | 3 | 409 |