Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039611.1 | NP_001034700.1 | 5 | 579 | ||
XM_006230291.2 | XP_006230353.1 | 4 | 421 | |||
XM_006230292.2 | XP_006230354.1 | 4 | 991 | |||
XM_006230293.2 | XP_006230355.1 | 4 | 501 | |||
GenBank mRNA | BC099757.1 | - | 5 | 579 | 5623851 |