Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039208.2 | NP_001034297.2 | 5 | 971 | ||
GenBank mRNA | BC110048.1 | - | 5 | 956 | 7315879 | |
BU671623.1 | - | 2 | 154 | |||
CA339235.1 | - | 5 | 544 |