Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130097.2 | NP_001123569.2 | 19 | 2562 | ||
XM_006230301.2 | XP_006230363.1 | 18 | 3600 | |||
XM_006230303.2 | XP_006230365.1 | 19 | 3488 | |||
XM_006230304.2 | XP_006230366.1 | 18 | 3600 | |||
XM_006230305.2 | XP_006230367.1 | 19 | 3506 | |||
XM_006230306.2 | XP_006230368.1 | 19 | 3487 | |||
XM_008759895.1 | XP_008758117.1 | 18 | 3600 | |||
XM_008759896.1 | XP_008758118.1 | 18 | 3582 | |||
XM_008759897.1 | XP_008758119.1 | 18 | 2539 | |||
XM_008759898.1 | XP_008758120.1 | 19 | 3506 | |||
XM_008759899.1 | XP_008758121.1 | 19 | 3488 | |||
XM_008759900.1 | XP_008758122.1 | 18 | 2887 | |||
XM_008759901.1 | XP_008758123.1 | 18 | 2838 | |||
GenBank mRNA | BC167761.1 | - | 19 | 2562 | 7442195 |