Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177599.1 | NP_001171070.1 | 4 | 401 | ||
NM_003848.3 | NP_003839.2 | 4 | 401 | |||
XM_017007415.1 | XP_016862904.1 | 4 | 397 | |||
XM_017007416.1 | XP_016862905.1 | 4 | 397 | |||
XM_017007417.1 | XP_016862906.1 | 4 | 397 | |||
XM_017007418.1 | XP_016862907.1 | 4 | 397 | |||
XM_017007419.1 | XP_016862908.1 | 4 | 397 | |||
XM_017007420.1 | XP_016862909.1 | 4 | 397 | |||
GenBank mRNA | AF058954.1 | - | 3 | 289 | ||
AK294598.1 | - | 4 | 394 | |||
AK310527.1 | - | 4 | 397 | |||
AL050226.1 | - | 1 | 64 | |||
BC007716.2 | - | 4 | 364 | 4303307 | ||
BC019868.1 | - | 1 | 40 | |||
BC035149.1 | - | 3 | 163 | |||
BC047024.1 | - | 5 | 371 | 5415743 | ||
BC068602.1 | - | 4 | 402 | 5274341 |