Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079894.1 | NP_001073363.1 | 6 | 545 | ||
XM_006230409.2 | XP_006230471.1 | 6 | 593 | |||
XM_006230412.2 | XP_006230474.1 | 6 | 594 | |||
XM_006230413.2 | XP_006230475.1 | 6 | 593 | |||
XM_006230415.2 | XP_006230477.1 | 6 | 593 | |||
XM_008759907.1 | XP_008758129.1 | 6 | 668 | |||
XM_008759908.1 | XP_008758130.1 | 6 | 989 | |||
XM_008759909.1 | XP_008758131.1 | 6 | 592 | |||
XM_008759910.1 | XP_008758132.1 | 3 | 232 | |||
GenBank mRNA | BC128752.1 | - | 6 | 545 | 7100072 |