Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177599.1 | NP_001171070.1 | 9 | 1060 | ||
NM_003848.3 | NP_003839.2 | 9 | 1060 | |||
XM_017007415.1 | XP_016862904.1 | 9 | 1056 | |||
XM_017007416.1 | XP_016862905.1 | 9 | 1056 | |||
XM_017007417.1 | XP_016862906.1 | 9 | 1056 | |||
XM_017007418.1 | XP_016862907.1 | 9 | 1056 | |||
XM_017007419.1 | XP_016862908.1 | 9 | 1056 | |||
XM_017007420.1 | XP_016862909.1 | 9 | 1056 | |||
GenBank mRNA | AF058954.1 | - | 8 | 948 | ||
AF131748.1 | - | 3 | 333 | |||
AK294598.1 | - | 9 | 1053 | |||
AK310527.1 | - | 9 | 1056 | |||
BC007716.2 | - | 9 | 1023 | 4303307 | ||
BC019868.1 | - | 6 | 699 | |||
BC035149.1 | - | 8 | 822 | |||
BC047024.1 | - | 10 | 1030 | 5415743 | ||
BC068602.1 | - | 9 | 1061 | 5274341 |