Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108508.1 | NP_001101978.1 | 11 | 1511 | ||
XM_006230562.1 | XP_006230624.1 | 14 | 1641 | |||
GenBank mRNA | BC166805.1 | - | 14 | 1421 | 7939004 |