Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039031.1 | NP_001034120.1 | 11 | 1231 | ||
XM_006231056.2 | XP_006231118.1 | 11 | 1117 | |||
XM_006231057.2 | XP_006231119.2 | 11 | 1400 | |||
GenBank mRNA | BC098925.1 | - | 11 | 1231 | 7112371 |