Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099402.1 | NP_001092872.1 | 5 | 591 | ||
XM_005268154.4 | XP_005268211.1 | 5 | 716 | |||
XM_011537275.2 | XP_011535577.1 | 5 | 888 | |||
GenBank mRNA | AB209373.1 | - | 2 | 1710 | ||
AK311075.1 | - | 5 | 569 | |||
BC015935.1 | - | 5 | 570 | 3907416 | ||
BQ432222.1 | - | 2 | 167 | |||
BT006950.1 | - | 4 | 461 |