Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212508.2 | NP_997673.1 | 4 | 793 | ||
XM_008772732.1 | XP_008770954.1 | 4 | 1916 | |||
GenBank mRNA | BC098637.1 | - | 4 | 945 | ||
FQ220138.1 | - | 3 | 574 |