Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BANF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143985.1 | NP_001137457.1 | 3 | 602 | ||
NM_003860.3 | NP_003851.1 | 3 | 659 | |||
XM_017018514.1 | XP_016874003.1 | 3 | 652 | |||
XM_017018515.1 | XP_016874004.1 | 3 | 192 | |||
GenBank mRNA | AF044773.1 | - | 3 | 255 | ||
AF068235.1 | - | 3 | 658 | |||
AF070447.1 | - | 2 | 151 | |||
AK311961.1 | - | 3 | 226 | |||
BC005942.1 | - | 3 | 238 | 4074168 | ||
BC107702.1 | - | 3 | 335 | 6495215 | ||
BU940708.1 | - | 3 | 419 | |||
CR542140.1 | - | 2 | 151 |