Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108535.1 | NP_001102005.1 | 22 | 3219 | ||
XM_006256462.1 | XP_006256524.1 | 22 | 3192 | |||
XM_006256463.1 | XP_006256525.1 | 22 | 3196 | |||
XM_006256464.2 | XP_006256526.1 | 21 | 3007 | |||
XM_008772893.1 | XP_008771115.1 | 20 | 4211 | |||
XM_008772894.1 | XP_008771116.1 | 22 | 3190 | |||
XM_008772895.1 | XP_008771117.1 | 20 | 4183 | |||
XM_008772896.1 | XP_008771118.1 | 19 | 4025 | |||
XM_008772897.1 | XP_008771119.1 | 18 | 2631 |