Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256500.2 | XP_006256562.1 | 14 | 2642 | ||
XM_008773011.1 | XP_008771233.1 | 16 | 2831 | |||
XM_008773013.1 | XP_008771235.1 | 15 | 2684 | |||
XM_008773015.1 | XP_008771237.1 | 17 | 2974 | |||
XM_008773016.1 | XP_008771238.1 | 16 | 2932 |