Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108538.1 | NP_001102008.1 | 6, 7 | 1009 | ||
XM_008773000.1 | XP_008771222.1 | 8, 9 | 979 | |||
GenBank mRNA | BC166724.1 | - | 8, 9 | 940 | 7933637 |