Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008372.1 | NP_001008373.1 | 10 | 1333 | ||
XM_006231776.2 | XP_006231838.1 | 10 | 1323 | |||
XM_006231778.2 | XP_006231840.1 | 10 | 1360 | |||
XM_008760639.1 | XP_008758861.1 | 11 | 1436 | |||
GenBank mRNA | BC085771.1 | - | 10 | 1333 | 7191068 | |
FQ211725.1 | - | 10 | 1352 |