Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srek1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008373.1 | NP_001008374.1 | 190 | |||
GenBank mRNA | AY236996.1 | - | 231 | |||
BC086553.1 | - | 190 | 7303262 |
Gene Symbol: LOC100362980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242128.2 | XP_006242190.1 | 1 | 543 | ||
XM_008765805.1 | XP_008764027.1 | 1 | 540 |