Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srek1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008373.1 | NP_001008374.1 | 166 | |||
GenBank mRNA | AY236996.1 | - | 207 | |||
BC086553.1 | - | 166 | 7303262 |