Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047909.1 | NP_001041374.1 | 4 | 504 | ||
GenBank mRNA | AY383686.1 | - | 4 | 504 |