Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl22l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108548.1 | NP_001102018.1 | 4 | 346 | ||
GenBank mRNA | BC167114.1 | - | 4 | 263 | 6919691 | |
FQ210535.1 | - | 4 | 262 | |||
FQ221224.1 | - | 4 | 262 | |||
FQ221258.1 | - | 4 | 260 | |||
FQ221490.1 | - | 4 | 262 | |||
FQ221726.1 | - | 4 | 262 | |||
FQ221812.1 | - | 4 | 262 | |||
FQ221852.1 | - | 4 | 325 | |||
FQ221904.1 | - | 4 | 263 | |||
FQ223163.1 | - | 4 | 262 | |||
FQ223463.1 | - | 4 | 262 | |||
FQ223753.1 | - | 4 | 261 | |||
FQ228314.1 | - | 4 | 262 | |||
FQ228345.1 | - | 4 | 262 | |||
FQ229046.1 | - | 4 | 262 |