Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014172.1 | NP_001014194.1 | 8 | 890 | ||
XM_008761055.1 | XP_008759277.1 | 8 | 880 | |||
GenBank mRNA | BC087665.1 | - | 8 | 890 | 7317835 |