Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1565775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342277.6 | XP_342278.5 | 10 | 1107 | ||
GenBank mRNA | BC092205.1 | - | 10 | 1060 | ||
BC103646.1 | - | 10 | 1085 |