Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324423.1 | NP_001311352.1 | 3 | 544 | ||
NM_004664.3 | NP_004655.1 | 4 | 663 | |||
NR_136887.1 | 4 | 621 | ||||
NR_136888.1 | 5 | 740 | ||||
XM_011538928.2 | XP_011537230.1 | 4 | 657 | |||
GenBank mRNA | AF028826.1 | - | 3 | 379 | ||
AF087693.1 | - | 4 | 632 | |||
AF173081.1 | - | 4 | 663 | |||
AK309256.1 | - | 4 | 452 | |||
AK315321.1 | - | 4 | 507 | |||
BC062332.1 | - | 5 | 605 | |||
BC099921.1 | - | 4 | 473 | 6729473 | ||
BC118609.1 | - | 4 | 607 | 40109202 | ||
BC122561.1 | - | 4 | 607 | 40109204 |